This item is now discontinued. Please browse similar items
Extech MG300-MTK Motor/Drive Troubleshooting Kit 20046-41
Representative image only.
Extech Mfr # MG300-MTK – Item # WZ-20046-41
Combines a 1000 V insulation tester with a full-featured True RMS MultiMeter in a single, compact instrument
  • Wireless USB interface transmits measurement data to a PC
  • Backlit super large triple display
  • 125 V, 250 V, 500 V, and 1000 V test voltages
  • Insulation resistance from 0.001 to 4000MΩ
  • Auto discharge of capacitive voltage
  • Lock power on function for hands-free operation
  • Data hold to freeze displayed reading
  • Min/Max and relative mode
  • Auto power off
  • Waterproof (IP67) rugged double-molded design
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMotor/Drive Troubleshooting Kit 20046-41
  • Warranty1 Year
1 Year WarrantyIP 67
Key Features
  • Wireless USB interface transmits measurement data to a PC
  • Backlit super large triple display
  • 125 V, 250 V, 500 V, and 1000 V test voltages
  • Insulation resistance from 0.001 to 4000MΩ
  • Auto discharge of capacitive voltage
  • Lock power on function for hands-free operation
  • Data hold to freeze displayed reading
  • Min/Max and relative mode
  • Auto power off
  • Waterproof (IP67) rugged double-molded design
  • 1 year warranty
More About this Item
MG300-MTK Motor & Drive Troubleshooting Kit includes:
MG300 True RMS 1000 V Multinmeter/insulation tester
480403 Motor rotation and three-phase tester
MA430T 400 A True RMS AC clamp meter with noncontact voltage