This item is now discontinued. Please browse similar items

Extech FS881 DMM Fuse Kit (1) 1A/600V and (1) 10A/600V Fuse.

Representative image only.
Extech Mfr # FS881 – Item # WZ-95001-96
Optimize your meter
  • Choose from accessories for various Extech meters

SPECS & DESCRIPTION

Specifications & Description

  • Compatible WithMM560, ML560 and MP560 Multimeters
  • Length (in)2
  • Height (in)3.2968
  • Width (in)0.4062
  • DescriptionDMM Fuse Kit (1) 1A/600V and (1) 10A/600V Fuse.

Key Features

  • Choose from accessories for various Extech meters

More About this Item

Convenient parts, carrying cases, and accessories for your Extech meters.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat