This item is now discontinued. Please browse similar items
Extech EX720 True RMS AC Clamp Meter, 800 Amps
Representative image only.
Extech Mfr # EX720 – Item # WZ-20005-37
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • AC Max (amps)800
  • Safety RatingCAT III 600V, EN 61010-1
  • DescriptionTrue RMS AC Clamp Meter, 800 Amps
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Functions include Inrush, Capacitance and Frequency
FEATURES
  • Models with AC/DC Current, AC/DC Voltage, Resistance, Frequency, Capacitance, Temperature, Diode and Continuity
  • 1ms Fast Peak Hold captures Inrush currents and transients
  • True RMS models
  • Double molded housing for better grip and one-handed operation
  • Large LCD display with bright backlighting
  • Data hold "freezes" data in display
  • 1.2" (30mm) circular jaw opening for large cables (500MCM)
  • Auto power off
  • Complete with test leads, 9V battery and Type K bead wire probe (EH720, EH730) and carrying case