This item is now discontinued. Please browse similar items
Extech ETK30 Electrical Test Kit with AC Clamp Meter
Representative image only.
Extech Mfr # ETK30 – Item # WZ-20046-48
Combines tools for testing for installation and repair of electrical systems and HVAC equipment
  • True RMS autoranging multimeter measures AC/DC voltage (600 V), AC/DC current (10 A with resolution to 0.1 µA), and resistance
  • 400 A AC clamp meter includes built-in noncontact voltage detector with LED alert
  • Clamp meter has jaw size of 1.2” (30 mm) for conductors up to 350 MCM
  • Both meters feature continuity/diode test, data hold, and max hold
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionElectrical Test Kit with AC Clamp Meter
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • True RMS autoranging multimeter measures AC/DC voltage (600 V), AC/DC current (10 A with resolution to 0.1 µA), and resistance
  • 400 A AC clamp meter includes built-in noncontact voltage detector with LED alert
  • Clamp meter has jaw size of 1.2” (30 mm) for conductors up to 350 MCM
  • Both meters feature continuity/diode test, data hold, and max hold