This item is now discontinued. Please browse similar items
Extech ET60 Voltage/Continuity Tester
Representative image only.
Extech Mfr # ET60 – Item # WZ-20000-87
Detect voltage quickly and safely
  • Seven-range LED indicators display AC and DC voltage range
  • Non-contact voltage detection range of 90 to 1000 V AC
  • AC frequency bandwidth of 45 to 66 Hz
  • Vibration alert when voltage of >85 V is detected
  • Double-molded housing with CAT III-600V rating
  • Continuity tester with audible and visual alert
  • Built-in test leads
  • Built-in flashlight for probing in dimly lit areas
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Max (Volts)220
  • AC Max (Volts)600
  • Safety RatingCAT III-600 V
  • PowerTwo AAA batteries
  • DescriptionVoltage/Continuity Tester
  • Warranty1 Year
1 Year WarrantyETL Listed
Key Features
  • Seven-range LED indicators display AC and DC voltage range
  • Non-contact voltage detection range of 90 to 1000 V AC
  • AC frequency bandwidth of 45 to 66 Hz
  • Vibration alert when voltage of >85 V is detected
  • Double-molded housing with CAT III-600V rating
  • Continuity tester with audible and visual alert
  • Built-in test leads
  • Built-in flashlight for probing in dimly lit areas
  • 1 year warranty
More About this Item
Safely detect the presence of voltage quickly and easily with the ET series of electrical testers. Visible and audible alarms notify you if live voltage is nearby and should be avoided or powered down. Choose from a wide selection of compact voltage and continuity testers for use in residential, commercial, and industrial applications.