This item is now discontinued. Please browse similar items
Extech ET28B Four-Range Voltage Tester
Representative image only.
Extech Mfr # ET28B – Item # WZ-20000-84
Detect voltage quickly and safely
  • Detects voltage from 120 to 480 V AC and 120 to 240 V DC
  • Neon light indicator flashes when voltage is detected
  • Test lead clip offers easy one-handed operation
  • Insulated housing provides added safety
  • Built-in test leads with removable protective caps and 29” (73.6 cm) cable
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Min (Volts)6
  • DC Max (Volts)240
  • AC Min (Volts)6
  • AC Max (Volts)480
  • DescriptionFour-Range AC Voltage Tester
  • Warranty1 Year
1 Year WarrantyETL Listed
Key Features
  • Detects voltage from 120 to 480 V AC and 120 to 240 V DC
  • Neon light indicator flashes when voltage is detected
  • Test lead clip offers easy one-handed operation
  • Insulated housing provides added safety
  • Built-in test leads with removable protective caps and 29” (73.6 cm) cable
  • 1 year warranty
More About this Item
Safely detect the presence of voltage quickly and easily with the ET series of electrical testers. Visible and audible alarms notify you if live voltage is nearby and should be avoided or powered down. Choose from a wide selection of compact voltage and continuity testers for use in residential, commercial, and industrial applications.