This item is now discontinued. Please browse similar items
Extech ET23B Low Voltage Tester
Representative image only.
Extech Mfr # ET23B – Item # WZ-20000-82
Detect voltage quickly and safely
  • Detects low voltage from 5 to 50 V AC/DC
  • Neon light indicator flashes when voltage is detected
  • Insulated housing provides added safety
  • Built-in pocket clip keeps unit secure and readily available
  • Built-in test leads
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Min (Volts)5
  • DC Max (Volts)50
  • AC Min (Volts)5
  • AC Max (Volts)50
  • DescriptionLow Voltage Tester
  • Warranty1 Year
1 Year WarrantyETL Listed
Key Features
  • Detects low voltage from 5 to 50 V AC/DC
  • Neon light indicator flashes when voltage is detected
  • Insulated housing provides added safety
  • Built-in pocket clip keeps unit secure and readily available
  • Built-in test leads
  • 1 year warranty
More About this Item
Safely detect the presence of voltage quickly and easily with the ET series of electrical testers. Visible and audible alarms notify you if live voltage is nearby and should be avoided or powered down. Choose from a wide selection of compact voltage and continuity testers for use in residential, commercial, and industrial applications.