This item is now discontinued. View the direct replacement: WZ-20034-53 or browse similar items
Extech DV24 Voltage Detector with Flashlight, 50 to 1000 VAC
Representative image only.
Extech Mfr # DV24 – Item # WZ-20034-52
  • For use on 50/60Hz circuits
  • Rugged double-molded housing
  • Tip fits into outlets or against wire insulation testing
  • Audible and visual voltage indication
  • Built-in bright flashlight with On/Off button
  • Compact size with pocket clip
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Min (Volts)50
  • DC Max (Volts)1000
  • AC Min (Volts)50
  • AC Max (Volts)1000
  • Safety RatingCAT IV 1000V, IEC 1010-1
  • DescriptionVoltage Detector with Flashlight, 50 to 1000 VAC
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • For use on 50/60Hz circuits
  • Rugged double-molded housing
  • Tip fits into outlets or against wire insulation testing
  • Audible and visual voltage indication
  • Built-in bright flashlight with On/Off button
  • Compact size with pocket clip