This item is now discontinued. Please browse similar items
Extech DCP60-220 DC Switching Power Supply, 600 W, 220 VAC
Representative image only.
Extech Mfr # DCP60-220 – Item # WZ-20046-47
Excellent for bench technicians, engineers, and educational facilities
  • Three user-defined voltage and current presets provide additional flexibility
  • Dual-action rotary encoder control with push knob for easy fine and coarse tuning
  • Limit maximum current to 5 A through front panel auxiliary output and 10 A from the back panel
  • Remote control for output voltage/current and on/off
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Output Voltage60 VDC
  • Output Current10 A
  • Ripple and Noise≤ 5 mVrms, < 100 mVp-p
  • Power Rating (watts)600
  • Power (VAC)220
  • DescriptionDC Switching Power Supply, 600 W, 220 VAC
  • Warranty1 Year
1 Year Warranty
Key Features
  • Three user-defined voltage and current presets provide additional flexibility
  • Dual-action rotary encoder control with push knob for easy fine and coarse tuning
  • Limit maximum current to 5 A through front panel auxiliary output and 10 A from the back panel
  • Remote control for output voltage/current and on/off