This item is now discontinued. We recommend the following as suggested replacements: WZ-16101-18
Extech 412400 Multi-Type Temperarture Calibrator
Representative image only.
Extech Mfr # 412400 – Item # WZ-16120-11
Light, compact design is ideal for field use
  • Provides adjustable 0 to 24 mA and 0 to 10 V calibration source
  • Five preset calibration values enable faster calibrations
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMulti-Type Temperarture Calibrator
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Provides adjustable 0 to 24 mA and 0 to 10 V calibration source
  • Five preset calibration values enable faster calibrations
  • 1 year warranty
More About this Item
Use this calibrator to set up and perform calibrations of process devices such as transducers, indicators, recorders, controllers and more. Dual-function calibrator simulates (source mode) and displays (measure mode) current and voltage outputs. Unit offers high accuracy (0.025%) with 24 VDC that drives current loads up to 1000 Ω. The “flip-up” design is easy-to-use and the six-button keypad makes calibration set ups and tests quick and simple. The large LCD displays readings in mA, as a percentage of the 0 to 24 mA range (4 to 20 mA = 100.0%), or as mV in voltage ranges.