Extech 380360 Insulation Tester/Megohmmeter
Representative image only.
Extech Mfr # 380360 – Item # WZ-26836-00
Measures insulation resistance of AC/DC motors, cables, power tools, and more!
  • Safety power lock — conduct a three-minute test hands-free
£238.00GBP / Each
£285.60GBP (incl VAT)
£55.00 - £154.00GBP / Each
£66.00 - £184.80GBP (incl VAT)
for an additional
£154.00GBP / Each
£184.80GBP (incl VAT)
for an additional
£55.00GBP / Each
£66.00GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • AC Current Range (mA)1
  • AC Voltage Accuracy±3% of reading 5 digits
  • Min Resistance (M ohms)0
  • Max Resistance (M ohms)2000
  • Resistance Resolution0.1Ω
  • Humidity80% RH
  • Min Temperature (° F)32
  • Max Temperature (° F)104
  • Min Temperature (° C)0
  • Max Temperature (° C)40
  • Display3.5 digit LCD
  • Battery Type (VAC)Six AA batteries
  • Height (in)6 19/64
  • Width (in)3 51/64
  • Depth (in)2 19/64
  • DescriptionInsulation Tester/Megohmmeter
Includes
six AA batteries, heavy-duty 30" test leads, alligator clip, and carrying case with neck strap.
Key Features
  • Safety power lock — conduct a three-minute test hands-free
More About this Item
This megohmmeter offers three test ranges: 200 MΩ, 250 V; 200 MΩ, 500 V; and 2000 MΩ, 1000 V. Full function indication updates testing conditions. Meter measures 0 to 600 VAC and features auto-off, data hold, auto-zero adjust, and overload protection.
ACCESSORIES
Add Optional Accessories