This item is now discontinued. Please browse similar items
Excel Scientific ThermalSeal RT2RR™ Sealing Film for Raised-Rim ABI Style Real-Time PCR, Polyester, Non-Sterile, 50µm; 100/PK
Representative image only.
Excel Scientific ThermalSeal Mfr # TS-RT2RR-100 – Item # WZ-13024-78
Eliminates the need for heat-sealing devices or mats
  • Designed for PCR, cold storage, and real-time PCR
  • Certified free of DNase-, RNase-, and nucleic-acids
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Min Temperature (° C)-40
  • Max Temperature (° C)120
  • MaterialTransparent polyester
  • Width (mm)142.9
  • Length (mm)79.4
  • DescriptionThermal Cycling Film for qPCR, Polyester, 50um, Raised-Rim Plates; 100/Pk
  • Warranty3 year
3 Year Warranty
Key Features
  • Designed for PCR, cold storage, and real-time PCR
  • Certified free of DNase-, RNase-, and nucleic-acids
  • 3 year warranty
More About this Item
These heat-resistant, adhesive sealing films prevent evaporation and protect samples during thermal cycling applications.