Excel Scientific ThermalSeal RT™ Sealing Film for Flat Top Real-Time PCR, Polyester, Non-Sterile, 50µm; 100/PK
Representative image only.
Excel Scientific ThermalSeal Mfr # TS-RT2-100 – Item # WZ-13024-76
Eliminates the need for heat-sealing devices or mats
  • Designed for PCR, cold storage, and real-time PCR
  • Certified free of DNase-, RNase-, and nucleic-acids
  • 3 year warranty
£188.53GBP / Pkg of 100
£226.24GBP (incl VAT)
Cole-Parmer Sealing Mat for 96-Well Plates, 2.2 mL, EVA; 50/pk
£299.60GBP / Pkg of 50
£359.52GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Min Temperature (° C)-40
  • Max Temperature (° C)125
  • MaterialTransparent polyester
  • Width (mm)142.9
  • Length (mm)79.4
  • DescriptionThermal Cycling Film for qPCR, Polyester, 50um; 100/Pk
  • Warranty3 year
3 Year Warranty
Key Features
  • Designed for PCR, cold storage, and real-time PCR
  • Certified free of DNase-, RNase-, and nucleic-acids
  • 3 year warranty
More About this Item
These heat-resistant, adhesive sealing films prevent evaporation and protect samples during thermal cycling applications.
Transparent polyester film is an optically clear film with a strong ultra-smooth, non-absorbing, non-fluorescing adhesive for superior performance in real-time PCR applications.