This item is now discontinued. Please browse similar items
Excel Scientific ThermalSeal Sealing Film for PCR, Polypropylene, Non-Sterile, 50µm; 100/PK
Representative image only.
Excel Scientific ThermalSeal Mfr # 100-THER-PLT – Item # WZ-13024-12
Eliminates the need for heat-sealing devices or mats
  • Designed for PCR, cold storage, and real-time PCR
  • Certified free of DNase-, RNase-, and nucleic-acids
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Min Temperature (° C)-40
  • Max Temperature (° C)125
  • MaterialHeat-resistant polypropylene
  • Width (mm)135.1
  • Length (mm)79.4
  • DescriptionFilm for Thermal Cycling, Polypropylene, 50µm, 100/pk
Key Features
  • Designed for PCR, cold storage, and real-time PCR
  • Certified free of DNase-, RNase-, and nucleic-acids
More About this Item
These heat-resistant, adhesive sealing films prevent evaporation and protect samples during thermal cycling applications.
Heat-resistant polypropylene films (13024-12 and -14) offer sufficient sealing area for all PCR plates.