This item is now discontinued. Please browse similar items
Excel Scientific ThermalSeal A Advanced, non-sterile films, 100/cs
Representative image only.
Excel Scientific ThermalSeal Mfr # TSA-100 – Item # WZ-13024-58
Films will not rollback – allows for easier application
  • Film fits between rims of PCR plates — ensures better adhering
  • Heat resistant film can be used from temperatures of -40 to +125°C
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° C)-40
  • Max Temperature (° C)125
  • MaterialPolypropylene
  • Width (mm)135.5
  • Length (mm)77.8
  • DescriptionA Advanced, non-sterile films, 100/cs
Key Features
  • Film fits between rims of PCR plates — ensures better adhering
  • Heat resistant film can be used from temperatures of -40 to +125°C
More About this Item
These heat-resistant, adhesive sealing films prevent evaporation and protect samples during thermal cycling applications. Films are made from 50µm polypropylene and are certified DNase-, RNase-, and nucleic acid-free. Non-pierceable, offering sufficient and reliable sealing for all PCR plates. Removable end tabs provide easier positioning on plates.