This item is now discontinued. Please browse similar items
Excel Scientific SealPlate MiniStrip Films, Sterile; 200/Pk
Representative image only.
Excel Scientific SealPlate Mfr # SPS-2X8-50 – Item # WZ-13024-06
Prevents evaporation and protects sample during processing, storage, and shipment
  • Superior quality film and uniform, consistently applied adhesive
  • Two removable end tabs for easy and accurate positioning on microplate
  • Functional temperature range from –40°C to 120°C
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Plate TypeMiniStrips film
  • Min Temperature (° C)-40
  • Max Temperature (° C)120
  • MaterialPolyester
  • Width (mm)19
  • Length (mm)102.9
  • DescriptionMiniStrip Films, Sterile; 200/Pk
Sterile
Key Features
  • Superior quality film and uniform, consistently applied adhesive
  • Two removable end tabs for easy and accurate positioning on microplate
  • Functional temperature range from –40°C to 120°C