This item is now discontinued. Please browse similar items
Excel Scientific SealPlate MiniStrip Films, Non-Sterile; 200/Pk
Representative image only.
Excel Scientific SealPlate Mfr # SP-2X8-50 – Item # WZ-13024-04
Prevents evaporation and protects sample during processing, storage, and shipment
  • Superior quality film and uniform, consistently applied adhesive
  • Two removable end tabs for easy and accurate positioning on microplate
  • Functional temperature range from –40°C to 120°C
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Plate TypeMiniStrips film
  • Min Temperature (° C)-40
  • Max Temperature (° C)120
  • MaterialPolyester
  • Width (mm)19
  • Length (mm)102.9
  • DescriptionMiniStrip Films, Non-Sterile; 200/Pk
Key Features
  • Superior quality film and uniform, consistently applied adhesive
  • Two removable end tabs for easy and accurate positioning on microplate
  • Functional temperature range from –40°C to 120°C
More About this Item
MiniStrips Films are identical to the microplate films above, but are designed for sealing only one or two 8-well rows at a time on either strip-well plates or standard microplates whenever rows must be selectively protected or accessed. Also use as a secondary seal to cover films that have been pierced. Each film measures 19 x 103.2 mm with the two removable end tabs; 19 x 82.6 mm without tabs.