This item is now discontinued. Please browse similar items
Erlab Filter Saturation Detection Alarm Module
Representative image only.
Erlab Mfr # MOLECODE – Item # WZ-33520-99
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFilter Saturation Detection Alarm Module
More About this Item
Choose from Replacement Filters, Work Surfaces, Carts, and Benches
This module attaches to the fume hood control panel and alerts user to filter over-saturation or accidental spills that has saturated the filter and needs replacing. Installed before hood shipment.