Eppendorf 022638955 Adapter for Swing Bucket Rotor 02570-65, PCR 384-Well plate, 2 per pack
Representative image only.
Eppendorf Mfr # 022638955 – Item # WZ-02570-95
Adapters suitable for a wide range of applications
PRODUCT OPTIONS
£198.54GBP / Pkg of 2
£238.25GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Tube Capacity1 x 384-well PCR plate per adapter
  • DescriptionAdapter for Swing Bucket Rotor 02570-65, PCR 384-Well plate, 2 per pack
More About this Item
Choose from adapters suitable for use with your Eppendorf 5804 and 5810 Series Multi-Purpose Centrifuges.