Environmental Express SAMPLING KITS Snip & Pour 10mL MeOH, 20 pk
Representative image only.
Environmental Express Item # WZ-35201-02
Extend sample hold time up to 14 days
  • All reagents and vials are certified and meet the requirements of EPA Method 5035
  • Keep vials upright and organized with the included plastic rack
  • Simply take your sample and place it in the vial—no field weighing required
PRODUCT OPTIONS
£98.18GBP / Pkg of 1
£117.82GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Tests Per Kit20
  • DescriptionSnip & Pour 10mL MeOH, 20 pk
Key Features
  • All reagents and vials are certified and meet the requirements of EPA Method 5035
  • Keep vials upright and organized with the included plastic rack
  • Simply take your sample and place it in the vial—no field weighing required
More About this Item
These field sampling kits come with everything you need to obtain field samples for volatile organics in soil and waste testing. Convenient, economical, and easy-to-use—prelabeled vials, reagents, certifications, coring device, and instructions are included. All reagents and vials are certified with chromatograms available.