Environmental Express SAMPLING KITS SB Premium kit, 10 mL MeOH in PTFE Tube
Representative image only.
Environmental Express Item # WZ-35200-91
Extend sample hold time up to 14 days
  • All reagents and vials are certified and meet the requirements of EPA Method 5035
  • Keep vials upright and organized with the included plastic rack
  • Simply take your sample and place it in the vial—no field weighing required
PRODUCT OPTIONS
£17.00GBP / Each
£20.40GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • TestVolatile organics in soil and waste testing
  • Reagent PlatformEPA Method 5035
  • Tests Per Kit1
  • DescriptionSB Premium kit, 10 mL MeOH in PTFE Tube
Key Features
  • All reagents and vials are certified and meet the requirements of EPA Method 5035
  • Keep vials upright and organized with the included plastic rack
  • Simply take your sample and place it in the vial—no field weighing required
More About this Item
These field sampling kits come with everything you need to obtain field samples for volatile organics in soil and waste testing. Convenient, economical, and easy-to-use—prelabeled vials, reagents, certifications, coring device, and instructions are included. All reagents and vials are certified with chromatograms available.