This item is now discontinued. We recommend the following as suggested replacements: WZ-35204-84
Environmental Express E082000C Cubitainer, LDPE, 9.5 Liter; 36/Cs
Representative image only.
Environmental Express Item # WZ-35208-98
Collapsible containers nest for efficient storage
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)9500
  • Capacity (L)9.5
  • Capacity (oz)321
  • Capacity (Gal)2.5
  • MaterialLow Density Polyethyene (LDPE)
  • Bottle ColorNatural
  • AutoclavableNo
  • DescriptionCubitainer, LDPE, 9.5 Liter; 36/Cs
Key Features
  • Meets Level 3 classification for contaminant-free sample containers
  • Just unscrew cap and fill to expand to total volume
More About this Item
Cubitainer® containers meet EPA recommended guidelines for contamination-free sample containers, but do not ship with a certificate of analysis.