Environmental Express Disposable Sterile Sampling Vials with Thiosulfate, Hinged Snap-Caps, PP, 125 mL; 200/CS
Representative image only.
Environmental Express Item # WZ-06045-81
Sterile vials are designed for coliform water sampling
  • Comes with all the necessary certificates of sterility
  • Vials with Thiosulfate are capable of neutralizing 15 mg of residual chlorine
  • Locking hinged snap-cap and tamper evident seal keeps samples secure
  • Holds a total volume of 125 mL
  • Model number D3200
PRODUCT OPTIONS
£86.35GBP / Case of 200
£103.62GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)125
  • Tube MaterialPolypropylene
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeHinged/snap
  • Cap MaterialPolypropylene
  • Cap colorClear
  • SterileYes
  • Fill100 mL
  • Special FeaturesVial with Thiosulfate
  • DescriptionDisposable Sterile Sampling Vials with Thiosulfate, Hinged Snap-Caps, Polypropylene, 125 mL; 200/Cs
Sterile
Includes
Sodium thiosulfate and hinged snap-cap.
Key Features
  • Comes with all the necessary certificates of sterility
  • Vials with Thiosulfate are capable of neutralizing 15 mg of residual chlorine
  • Locking hinged snap-cap and tamper evident seal keeps samples secure
  • Holds a total volume of 125 mL
  • Model number D3200
More About this Item
These disposable vials are sterile and ideal for coliform water sampling and sample dichlorination applications. Each vial is marked at the 100 mL fill line. Vials with thiosulfate contain enough sodium thiosulfate to neutralize 15 mg of residual chlorine. When filled, the vials meet the 0.008% sodium thiosulfate concentration requirement for sample collection containers.
Two types of vials are available—polypropylene vial with hinged snap-caps and clear polycarbonate vial with screw-top cap.