Environmental Express Air Sampling Quartz Fiber Filters, 47 mm; 100/Pk

Representative image only.
Environmental Express Item # WZ-35209-46
Ideal for air particle monitoring and environmental testing
  • Laser cutting ensures a clean, fiber-free filter edge
  • Excellent temperature and chemical resistance

PRODUCT OPTIONS

£82.09GBP / Pkg of 100
£98.51GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • MaterialQuartz Microfiber
  • Diameter (mm)47
  • Diameter (cm)4.7
  • Membrane SupportNo
  • DescriptionAir Sampling Quartz Fiber Filters, 47 mm; 100/Pk

Key Features

  • Laser cutting ensures a clean, fiber-free filter edge
  • Excellent temperature and chemical resistance

More About this Item

Quality filters to meet your industrial hygiene applications with excellent air permeability with minimal pressure drop.

Available for sale in the United States, UK, and India only

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat