EMD Millipore SCNY00100 Steriflip-GP Filter, 100 um Nylon, Sterile; 25/Pk
Representative image only.
EMD Millipore Mfr # SCNY00100 – Item # WZ-29969-28
For vacuum filtering solutions in 50-mL centrifuge tubes
  • Feature high-quality membranes
  • Millipore Express® (PES) membranes are ideal for low protein binding and exhibit faster flow rates with viscous solutions
  • Durapore® PVDF membranes offer the lowest protein binding available and are ideal for scale-up applications
  • Nylon mesh membranes are best for large particle removal
  • Nylon mesh membranes are utilized for large particle removal
PRODUCT OPTIONS
£191.11GBP / Pkg of 25
£229.33GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Tube Size (mL)50
  • MembraneNylon
  • Max Sample Volume (mL)50
  • Pore Size (µm)100
  • DescriptionSteriflip-GP Filter, 100 um Nylon, Sterile; 25/Pk
Sterile
Key Features
  • Feature high-quality membranes
  • Millipore Express® (PES) membranes are ideal for low protein binding and exhibit faster flow rates with viscous solutions
  • Durapore® PVDF membranes offer the lowest protein binding available and are ideal for scale-up applications
  • Nylon mesh membranes are best for large particle removal
  • Nylon mesh membranes are utilized for large particle removal
More About this Item
To filter, attach the unit directly to the 50-mL centrifuge tube containing your sample, flip it, and apply a vacuum. The filtrate collects directly in the attached 50-mL tube eliminating sample transfers between tubes. Disposable devices are sterile and have a 50-mL process volume.
Tube holder is not included.