This item is now discontinued. Please browse similar items
EMD Millipore PLBC07610 Ultrafiltration Disc Membranes, 3k 76mm 10pk
Representative image only.
EMD Millipore Mfr # PLBC07610 – Item # WZ-29949-38
Ideal for stirred cells and micropartition system
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (mm)76
  • Nominal Molecular Weight Limit (Mwl) (Dalton)3000
  • Diameter (cm)7.6
  • DescriptionUltrafiltration Disc Membranes, 3k 76mm 10pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Tight microstructure ensures the highest possible retention with the lowest possible adsorption of protein, DNA, or other macromolecules
  • 1 year warranty
More About this Item
These high-recovery hydrophilic membranes are made of regenerated cellulose. Use when concentrating or desalting extremely dilute solutions or whenever your sample is hydrophobic.