This item is now discontinued. We recommend the following as suggested replacements: WZ-18100-23
Elma Tec Clean S1 Ultrasonic Cleaner Detergent, Corrosion Remover; 25 L
Representative image only.
Elma Mfr # 800 0160 – Item # WZ-59988-59
Biodegradable, concentrated cleaners with long shelf lives
  • Several formulas to suit various industrial cleaning applications
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)6.6
  • Volume (Liters)25
  • ApplicationsManual or ultrasonic cleaning. Industrial use.
  • DescriptionUltrasonic Cleaner Detergent, Corrosion Remover; 25 L
Key Features
  • Several formulas to suit various industrial cleaning applications
More About this Item
S1 solution is mildly acidic to remove oil, grease, oxide films, rust, lime, and verdigris on nonferrous metal, aluminum, stainless steel, plastics, and glass—brightens brass and copper alloys.