This item is now discontinued. We recommend the following as suggested replacements: WZ-18100-23
Elma Tec Clean A1 Ultrasonic Cleaner Detergent, Electronics; 25 L
Representative image only.
Elma Mfr # 800 0100 – Item # WZ-59988-61
Biodegradable, concentrated cleaners with long shelf lives
  • Several formulas to suit various industrial cleaning applications
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)6.6
  • Volume (Liters)25
  • ApplicationsManual or ultrasonic cleaning. Industrial use.
  • DescriptionUltrasonic Cleaner Detergent, Electronics; 25 L
Key Features
  • Several formulas to suit various industrial cleaning applications
More About this Item
A1 solution is mildly alkaline and removes leftover light oils, fluxing agents, fingerprints, and dust—ideal for circuit boards and other fine electro-mechanical components.