This item is now discontinued. We recommend the following as suggested replacements: WZ-18100-23
Elma Lab Clean N10 Ultrasonic Cleaner Detergent, Foam-Inhibited; 10 L
Representative image only.
Elma Mfr # 800 0077 – Item # WZ-59988-72
Biodegradable, concentrated cleaners with long shelf lives
  • Solutions specifically made for labware
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)2.6
  • Volume (Liters)10
  • ApplicationsManual or ultrasonic cleaning. Lab use.
  • DescriptionUltrasonic Cleaner Detergent, Foam-Inhibited; 10 L
Key Features
  • Solutions specifically made for labware
More About this Item
N10 is a neutral, foam-inhibited cleaner for the removal of gentle emulsions, light greases and oils, fingerprints, and dust from most surfaces. Note: Test magnesium alloys before use.