This item is now discontinued. We recommend the following as suggested replacements: WZ-18100-23
Elma Lab Clean A20sf Ultrasonic Cleaner Detergent, Surfactant-Free; 25 L
Representative image only.
Elma Mfr # 800 0058 – Item # WZ-59988-69
Biodegradable, concentrated cleaners with long shelf lives
  • Solutions specifically made for labware
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)6.6
  • Volume (Liters)25
  • ApplicationsManual or ultrasonic cleaning. Lab use.
  • DescriptionUltrasonic Cleaner Detergent, Surfactant-Free; 25 L
Key Features
  • Solutions specifically made for labware
More About this Item
A20sf is a surfactant-free solution with ammonia—for cleaning of lab glassware especially for calibrated and volumetric glassware such as burettes, cylinders, and pipettes. Also suitable for plastics, stainless steel, and ceramics. Not intended for aluminum, magnesium, and other light-metal alloys.