This item is now discontinued. We recommend the following as suggested replacements: WZ-18100-23
Elma Clean 260 D&S Ultrasonic Cleaner Detergent, Gentle; 10 L
Representative image only.
Elma Mfr # 800 0073 – Item # WZ-59988-64
Biodegradable, concentrated cleaners with long shelf lives
  • Universal cleaner for lab and industrial use
SPECS & DESCRIPTION
Specifications & Description
  • FormatLiquid
  • Volume (gallons)2.6
  • Volume (Liters)10
  • ApplicationsManual or ultrasonic cleaning. Industrial or lab use.
  • DescriptionUltrasonic Cleaner Detergent, Gentle; 10 L
Key Features
  • Universal cleaner for lab and industrial use
More About this Item
This universal cleaner removes deposited lime soaps and inhibits re-deposit, grease, oils, fingerprints, and dust. Neutral, foam-inhibited, concentrated cleaning solution is perfect for most hard surfaces in lab and industrial applications. Note: Test magnesium alloys before use.