This item is now discontinued. Please browse similar items
Edwards C105-12-368 Blank Flange Vacuum Pump Fitting, Aluminum, NW 16; 1/Pk
Representative image only.
Edwards Mfr # C105-12-368 – Item # WZ-31400-00
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialAluminum
  • Connection TypeBlank Vacuum Flange NW 16
  • Flange Size (Port 1)NW 16
  • DescriptionBlank Flange Vacuum Pump Fitting, Aluminum, NW 16; 1/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Choose from chemically resistant 316 stainless steel (SS) or economical aluminum.
A blank flange will seal an outlet not in use. Elbow fittings provide a 90°> bend. Tee and cross fittings let you connect two or three lines to your pump. Order clamps and either co-seals or centering rings below.