Eagle 4510X Flammable Liquid Safety Cabinet, Self Closing, 2 Doors, 45 Gal

Representative image only.
Eagle Mfr # 4510X – Item # WZ-09418-27
Adjustable steel shelves hold up to 350 lb
  • Four adjustable galvanized leveling legs
  • Trilingual warning labels
  • Raised leakproof sill
  • Meeta NFPA Code 30 and OSHA requirements
  • 1 year warranty

PRODUCT OPTIONS

£1,848.35GBP / Each
£2,218.02GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Material18-gauge steel
  • Capacity (Liters)170.3
  • Capacity (Gal)45
  • Number of Shelves2
  • Width (in)43
  • Height (in)65
  • Depth (InchShort)18
  • Width (cm)109.2
  • Height (cm)165.1
  • Depth (cm)45.7
  • ColorYellow
  • Self-ClosingYes
  • ComplianceOSHA, NFPA Code 30
  • Shipping Weight (lbs)435
  • DescriptionFlammable Storage Cabinet, Self-Closing Doors, 45 Gallon
  • Warranty1 Year
1 Year WarrantyFM Approved

Key Features

  • Four adjustable galvanized leveling legs
  • Trilingual warning labels
  • Raised leakproof sill
  • Meeta NFPA Code 30 and OSHA requirements
  • 1 year warranty

More About this Item

Flammable cabinets are constructed of a high gloss powder coated finish, double-walled, 18-gauge steel with 1½" (3.8 cm) insulating air barrier — ideal for safely storing flammable liquid containers up to 5 gallons in size. Cabinets feature dual vents, each have 2" (5.1 cm) threaded fittings and a fire baffle and cap, grounding attachments, red trilingual warning, 2" (5.1 cm) raised, leakproof door sill, 3-point latch system, and flush mounted door handle with key lock. A concealed self-close mechanism provides obstruction-free access to top shelf space.

Includes adjustable galvanized steel shelves which direct spills to back and bottom of leakproof sump and hold 350 lbs (158.8 kg) Uniform Distributed Load (UDL). Hazard label: FLAMMABLE-KEEP FIRE AWAY.

Fetrures self-closing doors feature a fusible link that is actuated at 160°F/71°C.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat