This item is now discontinued. Please browse similar items
Eagle 1665 Overpack Salvage/Secondary Drum, 65 gallon
Representative image only.
Eagle Mfr # 1665 – Item # WZ-86110-00
For secondary storage, containment, transport, or clean-up of 55-gallon drums or smaller containers
  • Passed a 3-PSI pressure test.
  • UN HM-181 approved
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Liters)246.1
  • Capacity (Gal)65
  • Height (in)39
  • Height (cm)99.1
  • ComplianceDOT 49 CFR-173.3(c); 3psi pressure test for Salvage Drum Compliance
  • Shipping Weight (lbs)53
  • DescriptionOverpack Salvage/Secondary Drum, 65 gallon
Key Features
  • Passed a 3-PSI pressure test.
  • UN HM-181 approved
More About this Item
The salvage drums are constructed of yellow blow-molded high-density polyethylene (HDPE) with UV inhibitors, are durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration.
Drums are available with a metal band with bolt closure or screw top lids.