Eagle 1661 Qverpack Salvage Plastic Drum Barrel, Screw-On Lid, 65 Gal
Representative image only.
Eagle Mfr # 1661 – Item # WZ-86110-06
For secondary storage, containment, transport, or clean-up of 55-gallon drums or smaller containers
  • Passed a 3-PSI pressure test.
  • UN HM-181 approved
  • 1 year warranty
PRODUCT OPTIONS
£216.86GBP / Each
£260.23GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Liters)246.1
  • Capacity (Gal)65
  • Height (in)39
  • Height (cm)99.1
  • ComplianceDOT 49 CFR-173.3(c); 3psi pressure test for Salvage Drum Compliance
  • Shipping Weight (lbs)48
  • DescriptionOverpack Salvage Drum, screw top, 65 gallon
  • Warranty1 Year
1 Year Warranty
Key Features
  • Passed a 3-PSI pressure test.
  • UN HM-181 approved
  • 1 year warranty
More About this Item
The salvage drums are constructed of yellow blow-molded high-density polyethylene (HDPE) with UV inhibitors, are durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration.
Drums are available with a metal band with bolt closure or screw top lids.