Eagle 1618 Single-Drum Tray Dolly with Four 3" Casters
Representative image only.
Eagle Mfr # 1618 – Item # WZ-86110-44
Keeps container away from spills during dispensing
PRODUCT OPTIONS
£207.28GBP / Each
£248.74GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Gal)10
  • Height (in)7
  • Height (cm)17.8
  • Shipping Weight (lbs)27
  • DescriptionSingle-Drum HDPE Spill Tray Dolly
More About this Item
The single-drum spill tray is ideal for use under drums where dispensing or filling takes place. The drip pan catches up to one gallon of oils, coolants, water, or other non-aggressive liquids. Constructed of high-density polyethylene (HDPE). For use with single-drum tray dolly, model 86110-44.