Dynasonics DTTSLP-020-N-000 Ultrasonic Flow Transducer for 2" ANSI Pipe, 8 to 420 GPM (30 to 1590 LPM)

Representative image only.
Dynasonics Mfr # DTTSLP-020-N-000 – Item # WZ-32617-36
No need to shut down the process for installation or maintenance due to clamp-on sensor design
  • Bidirectional flow measurement system
  • Large, easy-to-read digital display
  • Rate and total backlit display
  • 4 to 20 mA output
  • RS-485 Modbus network connection

PRODUCT OPTIONS

£612.00GBP / Each
£734.40GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±1% of reading or ±0.01 ft/sec (0.003 m/s), whichever is greater
  • Material HousingPowder-coated aluminum, PP, SS, polyurethane, nickel-plated steel mounting brackets
  • Wetted MaterialsCPVC, Ultem®, nylon, and PVC
  • Min Operating Temperature (° F)-40
  • Min Operating Temperature (° C)-40
  • Max Operating Temperature (° F)185
  • Max Operating Temperature (° C)85
  • Pipe Size2" ANSI
  • Fluid TypeWater
  • Output (mA)Yes
  • Output4 to 20 mA
  • Display Type2-line LCD, LED backlight. Top row: 7 segment 0.7"H. Botton row: 0.35"H
  • CommunicationsRS-485
  • Enclosure RatingIP65
  • DescriptionUltrasonic Flow Transducer for 2" ANSI Pipe, 8 to 420 GPM (30 to 1590 LPM)

Key Features

  • Bidirectional flow measurement system
  • Large, easy-to-read digital display
  • Rate and total backlit display
  • 4 to 20 mA output
  • RS-485 Modbus network connection
  • Remote totalizer reset
  • 0 to 1000 Hz rate pulse and dual alarm output
  • USB programming port with software included
  • Rugged, aluminum enclosure ensures a long service life in harsh environments
  • Totalizer options include forward, reverse, and net total
  • Use to measure clean liquids as well as those with small amounts of suspended solids or aeration

More About this Item

Order Flowmeter 32617-24 with your transducer for a complete system!
Ultrasonic flowmeters clamp onto the outside of pipes and do not contact the internal liquid. The technology has inherent advantages over alternate devices including low-cost installation, no pressure head loss, no moving parts to maintain or replace, no fluid compatibility issue, and a large, bidirectional measuring range that ensures reliable readings even at very low and high flow rates. Flowmeters are available in a variety of configurations — select a meter with features suitable to meet your particular application requirements.

REVIEWS

Q&A

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    Will this flow meter work with PVC, Carbon Steel, Stainless Steel, and Copper?
    Asked on 1/30/2017 by Gene Kempers from Sioux City, Iowa

    1 answer

    • STAFF EXPERT

      A:

      32617-36 is just the transducer. Will work with ANSI pipe; PVC, Carbon Steel, Stainless Steel. For copper pipe use part number 32617-48.

      Answered on 1/31/2017 by Jeff from Technical Support
Displaying question 1

Do you have a question about this product? 

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat