This item is now discontinued. Please browse similar items
Dynalon Pointed-Tip Autoclavable Heavy-Duty PTFE Forceps, 4" L; 1/Pk
Representative image only.
Dynalon Mfr # 316514-0000 – Item # WZ-06442-08
Totally inert and non-contaminating PTFE Teweezer Forceps
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)4
  • Overall Length (cm)10.16
  • MaterialPTFE
  • SterileNo
  • DescriptionPointed-Tip Autoclavable Heavy-Duty PTFE Forceps, 4\" L; 1/Pk
More About this Item
Chemical-resistant, noncontaminating PTFE forceps are ideal for use in all applications. Heat resistant up to 572°F (300°C) for short periods and up to 500°F (260°C) consistently.