This item is now discontinued. Please browse similar items
DWK Life Sciences (Wheaton) W990016-CP CompletePAK® Sterile Vial Kit, 10 mL, Omni Flex 3G Stopper, Red; 158/Cs
Representative image only.
DWK Life Sciences (Wheaton) Mfr # W991016-CP – Item # WZ-08918-75
Reduce scrap costs—equal number of vials, stoppers, and seals
  • Vial packs have sustained particulate cleaning, depryogenation, and sterilization
  • Ready to use—right out of the box
  • 18-month shelf life from date of processing
  • 10 mL vials, 20 mm omni flex 3G stoppers, and red seals
  • Omni flex 3G stoppers are made from an ultra pure bromobutyl formulation with an additional fluoropolymer coating for further chemical purity and even lower gas permeability
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)10
  • Tube MaterialGlass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeOmni Flex 3G, standard
  • Cap Diameter (mm)20
  • Cap colorRed
  • SterileYes
  • AutoclavableYes
  • DescriptionCompletePAK® Sterile Vial Kit, 10 mL, Omni Flex 3G Stopper, Red; 158/Cs
NonpyrogenicSterile
Includes
10 mL clear-glass sterile vials, 20 mm sterile Omni Flex 3G serum stoppers, sterile red seals; case of 158
Key Features
  • Vial packs have sustained particulate cleaning, depryogenation, and sterilization
  • Ready to use—right out of the box
  • 18-month shelf life from date of processing
  • 10 mL vials, 20 mm omni flex 3G stoppers, and red seals
  • Omni flex 3G stoppers are made from an ultra pure bromobutyl formulation with an additional fluoropolymer coating for further chemical purity and even lower gas permeability
More About this Item
CompletePAK® vial kits include everything you need, without scrap costs, and no difference in component lead times. These clear-glass vials come with stoppers and seals—choose the volume, cap size and type, and seal color to suit your needs.
These presterilized vial kits are ready to use right off the shelf. Processing includes particulate cleaning, depryogenation, and sterilization—ideal for drug research, freeze drying/lyophilization, aseptic filling, and large/small molecule packaging.