This item is now discontinued. Please browse similar items
DWK Life Sciences (Wheaton) W990006-CP CompletePAK® Sterile Vial Kit, 5 mL, Ultra Pure Stopper, Blue; 200/Cs
Representative image only.
DWK Life Sciences (Wheaton) Mfr # W991006-CP – Item # WZ-08918-60
Reduce scrap costs—equal number of vials, stoppers, and seals
  • Vial packs have sustained particulate cleaning, depryogenation, and sterilization
  • Ready to use—right out of the box
  • 18-month shelf life from date of processing
  • 5 mL vials, 20 mm ultra-pure stoppers, and blue seals
  • Ultra-pure stoppers are made from an ultra pure bromobutyl formulation with extremely high chemical purity and low gas permeability
  • Compatible with applications such as Water for Injection (WFI) where rubber extractables can cause problems
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)5
  • Tube MaterialGlass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeUltra pure, standard
  • Cap Diameter (mm)20
  • Cap colorBlue
  • SterileYes
  • AutoclavableYes
  • DescriptionCompletePAK® Sterile Vial Kit, 5 mL, Ultra Pure Stopper, Blue; 200/Cs
NonpyrogenicSterile
Includes
5 mL clear-glass sterile vials, 20 mm sterile Ultra Pure serum stoppers, sterile blue seals; case of 200
Key Features
  • Vial packs have sustained particulate cleaning, depryogenation, and sterilization
  • Ready to use—right out of the box
  • 18-month shelf life from date of processing
  • 5 mL vials, 20 mm ultra-pure stoppers, and blue seals
  • Ultra-pure stoppers are made from an ultra pure bromobutyl formulation with extremely high chemical purity and low gas permeability
  • Compatible with applications such as Water for Injection (WFI) where rubber extractables can cause problems
More About this Item
CompletePAK® vial kits include everything you need, without scrap costs, and no difference in component lead times. These clear-glass vials come with stoppers and seals—choose the volume, cap size and type, and seal color to suit your needs.
These presterilized vial kits are ready to use right off the shelf. Processing includes particulate cleaning, depryogenation, and sterilization—ideal for drug research, freeze drying/lyophilization, aseptic filling, and large/small molecule packaging.