This item is now discontinued. View the direct replacement: WZ-34800-24 or browse similar items

DWK Life Sciences (Wheaton) W216946 Amber Wide-Mouth Bottle, PTFE-Faced Foam-Lined PP Cap, 2 oz, 24/cs

Representative image only.
DWK Life Sciences (Wheaton) Mfr # W216946 – Item # WZ-34632-32
  • Made of Type III Soda-Lime Glass
Often bought with…
Cole-Parmer® Essentials Glass Erlenmeyer Flasks, Graduated, Heavy Rim
£13.20 - £49.47GBP / Pkg of 8
£15.84 - £59.36GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)60
  • Capacity (oz)2
  • MaterialSoda-Lime Glass
  • Cap MaterialPolyethylene Foam-backed PTFE-lined Polypropylene
  • Cap / Stopper Size33-400
  • Cap ColorWhite
  • AutoclavableYes
  • Outside Diameter (OD) (cm)4.2
  • Overall Height (cm)8.5
  • DescriptionAmber Wide-Mouth Bottle, PTFE-Faced Foam-Lined PP Cap, 2 oz, 24/cs

Key Features

  • Made of Type III Soda-Lime Glass

More About this Item

Use these round glass bottles to store dry powders, soil samples and specimens. Ideal for general lab use or field sampling needs. Bottles include tamper evident labels that provide assurance that the bottle hasn't been tampered with in transit. Hazard communication labels are also included to communicate necessary information on the bottle. Caps are attached to the bottle; labels are not.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat