This item is now discontinued. Please browse similar items
DWK Life Sciences (Kimble) Kimax Glass Tilting Dispenser, 5 mL; 1/ea
Representative image only.
DWK Life Sciences (Kimble) Mfr # 759300-0005 – Item # WZ-07899-15
Features tilted head to allow accurate measurement
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)5
  • Joint Size Number24/40
  • DescriptionGlass Tilting Dispenser, 5 mL; 1/ea
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Manufactured entirely of glass; dispenser features a fixed volume dispensing head secured with a head. Flasks tilts back to fill to diseired volume, forward to dispense. Clamp is included.