This item is now discontinued. Please browse similar items

DWK Life Sciences (Kimble) KC14950-25 Heavy-Duty Solution Bottle (Carboy) 9.5 L / 2.5 gal.

Representative image only.
DWK Life Sciences (Kimble) Mfr # KC14950-25 – Item # WZ-34523-29
Controls spills in the event of glassware breakage
  • Ideal for chemical storage
  • Autoclavable to120°C (250°F)
Often bought with…
Cole-Parmer® Essentials Glass Erlenmeyer Flasks, Graduated, Heavy Rim
£13.20 - £49.47GBP / Pkg of 8
£15.84 - £59.36GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (L)9.5
  • Capacity (Gal)2.5
  • MaterialGlass
  • Cap / Stopper Size#12
  • Bottle ColorClear
  • AutoclavableYes
  • Withstands High Temperature (° F)250
  • Outside Diameter (OD) (in)8 3/4
  • Outside Diameter (OD) (cm)22.2
  • Overall Height (in)15 1/2
  • Overall Height (cm)39.2
  • DescriptionHeavy-Duty Solution Bottle (Carboy) 9.5 L / 2.5 gal.

Key Features

  • Ideal for chemical storage
  • Autoclavable to120°C (250°F)

More About this Item

Ultra-clear safety-coated glassware helps stop and control spills in the event of glassware breakage. If a break does occur, the durable odor-free glassware will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. Glassware is ideal for chemical storage, transportation, and handling of hazardous chemicals, harmful biologicals, and acids. Glassware has a unique nonslip surface when wet or dry. Durable bottles are tooled to fit a #12 size stopper. Five gallon size is designed from Federal Spec DD-B-597. Autoclavable to120°C (250°F).



REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat