DWK Life Sciences (Kimble) CPMUG600 Beaker Mug, glass, 20 ounce, graduated, 1/pk
Representative image only.
DWK Life Sciences (Kimble) Mfr # CPMUG600 – Item # WZ-34502-99
Easily portable with durable handle
  • Constructed of durable borosilicate glass
£38.45GBP / Pkg of 1
£46.14GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)600
  • Capacity (oz)20
  • MaterialGlass
  • AutoclavableYes
  • DescriptionBeaker Mug, glass, 20 ounce, graduated, 1/pk
Key Features
  • Constructed of durable borosilicate glass
More About this Item
Large 20-oz beaker mug features a handle for easy carrying and a lip around the top to prevent dripping. Mugs are made of borosilicate glass and are graduated every 100 mL from 100 to 600 mL.