DWK Life Sciences (Kimble) 73750-13100 Disposable Culture Tubes, 13 x 100, with markings 1000/cs
Representative image only.
DWK Life Sciences (Kimble) Mfr # 73750-13100 – Item # WZ-34580-00
Ideal for general bacteriological and tissue culture applications
  • Tubes are offered with or without white markings
PRODUCT OPTIONS
£495.50GBP / Case of 1000
£594.60GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)8
  • MaterialGlass
  • Cap / Stopper Size13-415
  • Bottle ColorClear
  • AutoclavableYes
  • Label TypeWhite Write-On
  • Outside Diameter (OD) (cm)1.3
  • Overall Height (cm)10
Key Features
  • Tubes are offered with or without white markings
More About this Item
Manufactured of borosilicate glass, these disposable tubes are ideal for general bacteriological and tissue culture applications. Caps are sold separately.