DWK Life Sciences (Kimble) 20024-250 250 mL class B graduated cylinders
Representative image only.
DWK Life Sciences (Kimble) Mfr # 20024-250 – Item # WZ-34527-08
Calibrated “to deliver” these single metric scale graduated cylinders meet ASTM Specification E1272, Style 1, Class B requirements
  • Single metric scale
  • Limited Lifetime Warranty
PRODUCT OPTIONS
£603.97GBP / Case of 12
£724.76GBP (incl VAT)
£90.44 - £142.45GBP / Each
£108.53 - £170.94GBP (incl VAT)
for an additional
£90.44GBP / Each
£108.53GBP (incl VAT)
for an additional
£142.45GBP / Each
£170.94GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
Cole-Parmer Essentials Amber Cylinder, Class B, To Deliver, 250 mL
£67.52GBP / Each
£81.02GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)250
  • Height (in)12 63/64
  • Capacity (oz)8
  • Graduation Interval (mL)2
  • Tolerance (mL)±1.4
  • Overall Height (cm)32.9946
  • Overall Height (in)12 63/64
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Single metric scale
  • Limited Lifetime Warranty
More About this Item
Bumpers are included with 25- to 2000-mL cylinders. Hexagonal base on all cylinders increases stability; the 10- and 25-mL cylinders are comparatively shorter for even greater stability. The enlarged funnel-shaped top on the 10-mL cylinder makes filling easy.