This item is now discontinued. Please browse similar items
DWK Life Sciences (Kimble) 20024-1000 1000 mL class B graduated cylinders
Representative image only.
DWK Life Sciences (Kimble) Mfr # 20024-1000 – Item # WZ-34527-12
Calibrated “to deliver” these single metric scale graduated cylinders meet ASTM Specification E1272, Style 1, Class B requirements
  • Single metric scale
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)1000
  • Graduation Interval (mL)10
  • Tolerance (mL)±5.0
  • Description1000 mL class B graduated cylinders
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Single metric scale
  • Limited Lifetime Warranty
More About this Item
Bumpers are included with 25- to 2000-mL cylinders. Hexagonal base on all cylinders increases stability; the 10- and 25-mL cylinders are comparatively shorter for even greater stability. The enlarged funnel-shaped top on the 10-mL cylinder makes filling easy.