This item is now discontinued. Please browse similar items
DuPont Tyvek Lab Coat, SnapFront, Serged Seams, Large; 1/Pk
Representative image only.
DuPont Mfr # TY212SWHLG003000 – Item # WZ-86225-32
Protect against airborne contaminants
SPECS & DESCRIPTION
Specifications & Description
  • SizeLarge
  • MaterialPolyethylene(PE)
  • ColorWhite
  • SeamSerged
  • DescriptionLab Coat, SnapFront, Serged Seams, Large; 1/Pk
More About this Item
Rugged Tyvek® lab coats resists snags, tears, punctures, and abrasion. The nonwoven fabric is chemically and biologically inert, low linting, and static-free until laundered. White lab coats have serged seams, five-snap front, and a collar.