This item is now discontinued. Please browse similar items
DuPont PC143SWH00250B Coveralls, x-large, bulk packed
Representative image only.
DuPont Mfr # PC143SWHXL00250B – Item # WZ-33684-28
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyethylene(PE)
  • SizeX-Large
  • ColorWhite
  • SeamSerged
  • ComplianceISO Class 6, 7, 8 (formely Class 1000, 10000, 100000)
  • DescriptionCoveralls, x-large, bulk packed
More About this Item
Pro/Clean® Coveralls are made from a unique breathable composite material for critical environments. This fabric provides excellent particulate and light splash protection. Coveralls feature a zipper front, serged seams, and elastic wrists, ankles, and waist. White.
Compliance: ISO Class 6, 7, and 8 (formerly Class 1000, 10,000, and 100,000)