Drierite 13025 Nonindicating desiccant; 25-lb can
Representative image only.
Drierite Mfr # 13025 – Item # WZ-07193-30
Use in desiccators, drying columns, or drying tubes
  • Keeps samples dry and moisture free
  • 1 year warranty
PRODUCT OPTIONS
£255.13GBP / Each
£306.16GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNonindicating dessicant, 25 lb can
  • ColorWhite
  • DescriptionNonindicating desiccant; 25-lb can
  • Warranty1 year
1 Year Warranty
Key Features
  • Keeps samples dry and moisture free
  • 1 year warranty
More About this Item
Use Drierite desiccants (anhydrous calcium sulfate) in desiccators, drying columns, or drying tubes. Choose indicating or non-indicating desiccant — indicating desiccant starts blue, then turns pink when regeneration or renewal is necessary.